Assay Details
| Gene Symbol: Igfbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008048.3 | NP_032074.3 | 1 - 2 | 503 | 81 | |
| GenBank mRNA | AB012886.1 | - | 1 - 2 | 504 | 81 | |
| AK002277.1 | - | 1 - 2 | 499 | 81 | ||
| AK148749.1 | - | 1 - 2 | 500 | 81 | ||
| AK156315.1 | - | 1 - 2 | 505 | 81 | ||
| AW318969.1 | - | 1 - 2 | 498 | 81 | ||
| BC047202.1 | - | 1 - 2 | 415 | 81 | ||
| BC092538.1 | - | 1 - 2 | 499 | 30284911 | 81 | |
| L75822.2 | - | 1 - 2 | 482 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp7
Genomic Map