Assay Details
| Gene Symbol: Lnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159577.1 | NP_001153049.1 | 1 - 2 | 113 | 86 | |
| GenBank mRNA | AF034745.1 | - | 1 - 2 | 113 | 86 | |
| CA469116.1 | - | 1 - 2 | 150 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF034745.1, NM_001159577.1 transcripts.
Set Membership: |
|

Lnx1
Genomic Map