Assay Details
| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145920.3 | NP_666032.1 | 2 - 3 | 320 | 76 | |
| GenBank mRNA | AB083066.1 | - | 2 - 3 | 250 | 76 | |
| AK044977.1 | - | 2 - 3 | 191 | 76 | ||
| AK134298.1 | - | 2 - 3 | 303 | 76 | ||
| AK165051.1 | - | 2 - 3 | 205 | 76 | ||
| BC037473.1 | - | 2 - 3 | 301 | 4237472 | 76 | |
| BC064473.1 | - | 2 - 3 | 311 | 3156380 | 76 | |
| BY270131.1 | - | 2 - 3 | 321 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044977.1 transcripts.
Set Membership: |
|

Evc2
Genomic Map