Assay Details
| Gene Symbol: Pacrgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025755.3 | NP_080031.1 | 7 - 8 | 722 | 86 | |
| GenBank mRNA | AK014276.1 | - | 7 - 8 | 733 | 86 | |
| AK016966.1 | - | 7 - 8 | 771 | 86 | ||
| AK018101.1 | - | 7 - 8 | 731 | 86 | ||
| AK032498.1 | - | 6 - 7 | 2470 | 86 | ||
| BC018481.1 | - | 7 - 8 | 722 | 3482169 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pacrgl
Genomic Map