Assay Details
| Gene Symbol: Zfyve28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015039.1 | NP_001015039.1 | 1 - 2 | 227 | 79 | |
| GenBank mRNA | AK036334.1 | - | 1 - 2 | 230 | 79 | |
| AK129414.1 | - | 1 - 2 | 227 | 79 | ||
| BC139050.1 | - | 1 - 2 | 168 | 8862072 | 79 | |
| BC139051.1 | - | 1 - 2 | 168 | 8862073 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve28
Genomic Map