Assay Details
| Gene Symbol: Zfyve28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015039.1 | NP_001015039.1 | 4 - 5 | 702 | 58 | |
| GenBank mRNA | AK036334.1 | - | 4 - 5 | 705 | 58 | |
| AK129414.1 | - | 4 - 5 | 702 | 58 | ||
| AK159159.1 | - | 3 - 4 | 723 | 58 | ||
| BC139051.1 | - | 4 - 5 | 643 | 8862073 | 58 | |
More Information
Set Membership: |
|

Zfyve28
Genomic Map