Assay Details
| Gene Symbol: Zfyve28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015039.1 | NP_001015039.1 | 5 - 6 | 793 | 70 | |
| GenBank mRNA | AK036334.1 | - | 5 - 6 | 796 | 70 | |
| AK129414.1 | - | 5 - 6 | 793 | 70 | ||
| AK159159.1 | - | 4 - 5 | 814 | 70 | ||
| BC139050.1 | - | 5 - 6 | 734 | 8862072 | 70 | |
| BC139051.1 | - | 5 - 6 | 734 | 8862073 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve28
Genomic Map