Assay Details
| Gene Symbol: Cpeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177379.1 | NP_001170850.1 | 9 - 10 | 2570 | 88 | |
| NM_175937.3 | NP_787951.2 | 9 - 10 | 2636 | 88 | ||
| GenBank mRNA | AB100307.1 | - | 10 - 11 | 1269 | 88 | |
| AK076221.1 | - | 3 - 4 | 332 | 88 | ||
| BC107349.1 | - | 10 - 11 | 1246 | 40055453 | 88 | |
| BC107350.1 | - | 10 - 11 | 1246 | 40055458 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpeb2
Genomic Map