Assay Details
| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114660.2 | NP_001108132.1 | 6 - 7 | 1830 | 99 | |
| NM_001286437.1 | NP_001273366.1 | 6 - 7 | 1830 | 99 | ||
| GenBank mRNA | AK088353.1 | - | 6 - 7 | 1716 | 99 | |
| AK089011.1 | - | 6 - 7 | 1531 | 99 | ||
| AK089153.2 | - | 7 - 8 | 1847 | 99 | ||
| AK135666.1 | - | 6 - 7 | 1066 | 99 | ||
| AK140246.1 | - | 6 - 7 | 1831 | 99 | ||
| AK143615.1 | - | 6 - 7 | 1810 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scfd2
Genomic Map