Assay Details
| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114660.2 | NP_001108132.1 | 7 - 8 | 1965 | 95 | |
| GenBank mRNA | AK088353.1 | - | 7 - 8 | 1851 | 95 | |
| AK089153.2 | - | 8 - 9 | 1982 | 92 | ||
| AK135666.1 | - | 7 - 8 | 1201 | 95 | ||
| AK140246.1 | - | 7 - 8 | 1966 | 95 | ||
| AK143615.1 | - | 7 - 8 | 1945 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scfd2
Genomic Map