Assay Details
| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114660.2 | NP_001108132.1 | 8 - 9 | 2082 | 109 | |
| GenBank mRNA | AK088353.1 | - | 8 - 9 | 1968 | 109 | |
| AK135666.1 | - | 8 - 9 | 1318 | 109 | ||
| AK140246.1 | - | 8 - 9 | 2083 | 109 | ||
| AK143615.1 | - | 8 - 9 | 2062 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scfd2
Genomic Map