| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_001159573.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_001159573.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_001159574.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_001159574.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_024183.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
NM_024183.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006198.mm.1 |
TC0500000701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006199.mm.1 |
NM_001159573.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006199.mm.1 |
NM_001159573.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006199.mm.1 |
NM_024183.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006199.mm.1 |
NM_024183.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006199.mm.1 |
TC0500000701.mm.1 |
MTA TC |