Assay Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011914.2 | NP_036044.1 | 1 - 2 | 234 | 77 | |
| GenBank mRNA | AK019191.1 | - | 1 - 2 | 299 | 77 | |
| AK078760.1 | - | 1 - 2 | 234 | 77 | ||
| AK088384.1 | - | 1 - 2 | 250 | 77 | ||
| AK137949.1 | - | 1 - 2 | 227 | 77 | ||
| AK144173.1 | - | 248 | 77 | |||
| AK146185.1 | - | 1 - 2 | 286 | 77 | ||
| BC038003.1 | - | 1 - 2 | 235 | 4912719 | 77 | |
More Information
Set Membership: |
|

Nelfa
Genomic Map