Assay Details
| Gene Symbol: Ndufaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085493.2 | NP_001078962.1 | 5 - 6 | 611 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157939 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145689 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05155144 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145689 | - | - |
| Whole Mouse Genome G4122A | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150525 | NM_001085493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150525 | AK009547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150525 | BC048539.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22727966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03429042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03618383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03429619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03368005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03551057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03758277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03758283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04044935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03695735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03816653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03551059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03551055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03758280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03488119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03393788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03816647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03816651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03305261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03816655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03727703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03652251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03352281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22727949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03546909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10511504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511502 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429213_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511501 | - | - |
| Mouse Genome 430 2.0 Array | 1429213_at | - | - |
| Mouse Genome 430 2.0 Array | 1429213_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76947_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76947_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400018343.mm.1 | NM_001085493.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018343.mm.1 | TC0400002200.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429213_at | - | - |
| Mouse Expression Set 430 | 1429213_PM_at | - | - |
| Murine Genome U74v2 Set | 165986_at | - | - |
| Murine Genome U74v2 Set | 165986_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429213_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429213_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 631876 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656042 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2656042 | - | - |

Ndufaf6
Genomic Map