Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138758.1 | NP_620097.1 | 3 - 4 | 493 | 129 | |
| GenBank mRNA | AK052425.1 | - | 3 - 4 | 495 | 129 | |
| AK144627.1 | - | 3 - 4 | 489 | 129 | ||
| AK161724.1 | - | 3 - 4 | 460 | 129 | ||
| AY033513.1 | - | 2 - 3 | 331 | 129 | ||
| BC010495.1 | - | 3 - 4 | 493 | 4195680 | 129 | |
| BC115365.1 | - | 3 - 4 | 425 | 40059106 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmlhe
Genomic Map