Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138758.1 | NP_620097.1 | 6 - 7 | 1124 | 107 | |
| GenBank mRNA | AK144627.1 | - | 6 - 7 | 1120 | 107 | |
| AK161724.1 | - | 6 - 7 | 1091 | 107 | ||
| AY033513.1 | - | 5 - 6 | 962 | 107 | ||
| BC010495.1 | - | 6 - 7 | 1124 | 4195680 | 107 | |
| BC115365.1 | - | 6 - 7 | 1056 | 40059106 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmlhe
Genomic Map