Assay Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172988.4 | NP_766576.1 | 4 - 5 | 1176 | 85 | |
| NR_104378.1 | 4 - 5 | 1176 | 85 | |||
| NR_104379.1 | 4 - 5 | 1176 | 85 | |||
| GenBank mRNA | AK028786.1 | - | 4 - 5 | 1160 | 85 | |
| AK032391.1 | - | 4 - 5 | 1155 | 85 | ||
| AK051429.1 | - | 4 - 5 | 1127 | 85 | ||
| BC033265.1 | - | 4 - 5 | 1103 | 3983942 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl4
Genomic Map