Assay Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC117535.1 | - | 2 - 3 | 1115 | 40106181 | 117 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155a| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC117535.1 | - | 2 - 3 | 1115 | 40106181 | 117 |
Set Membership: |
|