Assay Details
| Gene Symbol: Pcid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178708.3 | NP_848823.2 | 7 - 8 | 999 | 66 | |
| GenBank mRNA | AK042945.1 | - | 7 - 8 | 489 | 66 | |
| AK045796.1 | - | 7 - 8 | 479 | 66 | ||
| AK048722.1 | - | 7 - 8 | 490 | 66 | ||
| AK082891.1 | - | 7 - 8 | 1065 | 66 | ||
| AK163697.1 | - | 7 - 8 | 486 | 66 | ||
| BC006047.1 | - | 7 - 8 | 488 | 66 | ||
| BC072621.1 | - | 4 - 5 | 933 | 66 | ||
| BC118533.1 | - | 7 - 8 | 999 | 40095328 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcid2
Genomic Map