Assay Details
| Gene Symbol: Pcid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178708.3 | NP_848823.2 | 1 - 2 | 571 | 76 | |
| GenBank mRNA | AK042945.1 | - | 1 - 2 | 61 | 76 | |
| AK045796.1 | - | 1 - 2 | 51 | 76 | ||
| AK048722.1 | - | 1 - 2 | 62 | 76 | ||
| AK082891.1 | - | 1 - 2 | 636 | 76 | ||
| AK163697.1 | - | 1 - 2 | 58 | 76 | ||
| BC072621.1 | - | 1 - 2 | 687 | 76 | ||
| BC118533.1 | - | 1 - 2 | 571 | 40095328 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082891.1 transcripts.
Set Membership: |
|

Pcid2
Genomic Map