Assay Details
| Gene Symbol: Pcid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178708.3 | NP_848823.2 | 9 - 10 | 1222 | 64 | |
| GenBank mRNA | AK045796.1 | - | 9 - 10 | 702 | 64 | |
| AK048722.1 | - | 9 - 10 | 713 | 64 | ||
| AK082891.1 | - | 9 - 10 | 1288 | 64 | ||
| AK163697.1 | - | 9 - 10 | 709 | 64 | ||
| BC006047.1 | - | 9 - 10 | 711 | 64 | ||
| BC072621.1 | - | 6 - 7 | 1156 | 64 | ||
| BC118533.1 | - | 9 - 10 | 1222 | 40095328 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcid2
Genomic Map