Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 12 - 13 | 2902 | 134 | |
| GenBank mRNA | BC057401.1 | - | 11 - 12 | 2811 | 5697758 | 134 |
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map