Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 5 - 6 | 2072 | 89 | |
| GenBank mRNA | AK089130.1 | - | 6 - 7 | 1626 | 89 | |
| AK089143.1 | - | 7 - 8 | 2423 | 89 | ||
| BU707304.1 | - | 1 - 2 | 92 | 89 | ||
| U07357.1 | - | 5 - 6 | 1612 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map