Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143671.1 | NP_001137143.1 | 1 - 2 | 213 | 107 | |
| NM_001143672.1 | NP_001137144.1 | 1 - 2 | 213 | 107 | ||
| GenBank mRNA | AK131817.1 | - | 1 - 2 | 213 | 107 | |
| BC089620.1 | - | 1 - 2 | 189 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143671.1, NM_001143672.1 transcripts.
Set Membership: |
|

Tmem255b
Genomic Map