Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143671.1 | NP_001137143.1 | 3 - 4 | 421 | 70 | |
| GenBank mRNA | AK131817.1 | - | 3 - 4 | 421 | 70 | |
More Information
Set Membership: |
|

Tmem255b
Genomic Map