Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143671.1 | NP_001137143.1 | 5 - 6 | 590 | 70 | |
| NM_001143672.1 | NP_001137144.1 | 4 - 5 | 527 | 70 | ||
| GenBank mRNA | AK131817.1 | - | 5 - 6 | 590 | 70 | |
| BC039763.1 | - | 3 - 4 | 354 | 70 | ||
| BC042836.1 | - | 3 - 4 | 343 | 70 | ||
| BC089620.1 | - | 4 - 5 | 502 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem255b
Genomic Map