Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143671.1 | NP_001137143.1 | 7 - 8 | 830 | 67 | |
| NM_001143672.1 | NP_001137144.1 | 6 - 7 | 767 | 67 | ||
| GenBank mRNA | AK131817.1 | - | 7 - 8 | 831 | 67 | |
| BC039763.1 | - | 5 - 6 | 594 | 67 | ||
| BC042836.1 | - | 5 - 6 | 583 | 67 | ||
| BC089620.1 | - | 6 - 7 | 742 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem255b
Genomic Map