Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143671.1 | NP_001137143.1 | 8 - 9 | 973 | 93 | |
| NM_001143672.1 | NP_001137144.1 | 7 - 8 | 910 | 93 | ||
| GenBank mRNA | AK131817.1 | - | 8 - 9 | 974 | 93 | |
| BC039763.1 | - | 6 - 7 | 737 | 93 | ||
| BC042836.1 | - | 6 - 7 | 726 | 93 | ||
| BC089620.1 | - | 7 - 8 | 885 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054989 | BC039763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054989 | BC042836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054989 | AK131817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054989 | BC089620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054989 | NM_001143671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108032 | BC039763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108032 | BC042836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108032 | AK131817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108032 | BC089620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108032 | NM_001143671.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11271 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108032 | BC039763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108032 | BC042836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108032 | AK131817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108032 | BC089620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108032 | NM_001143671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054989 | BC039763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054989 | BC042836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054989 | AK131817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054989 | BC089620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054989 | NM_001143671.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06691635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07235527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06997897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07235512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06879770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07103557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06879774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07195101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25668313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570392 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570388 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001203.mm.1 | NM_001143671.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001203.mm.1 | NM_001143671.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001203.mm.1 | NM_001143672.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001203.mm.1 | NM_001143672.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001203.mm.1 | TC0800000128.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2533140 | - | - |

Tmem255b
Genomic Map