Assay Details
| Gene Symbol: Gm15984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040684.1 | - | 2 - 3 | 287 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ligand of numb-protein X 1 | Lnx1 | Lnx | -8.8kb | Anti-sense | Chr. 5 - 74592447 - 74703042 on NCBI Build GRCm38 |
| Downstream | predicted gene 6116 | Gm6116 | EG619945 | 236.1kb | Sense | Chr. 5 - 74949194 - 74966368 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15984
Genomic Map