Assay Details
| Gene Symbol: Cytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081106.1 | NP_001074575.1 | 1 - 2 | 186 | 108 | |
| GenBank mRNA | BC063103.1 | - | 1 - 2 | 146 | 108 | |
| BC116224.1 | - | 1 - 2 | 160 | 40091368 | 108 | |
| BC116225.1 | - | 1 - 2 | 160 | 40091369 | 108 | |
| EF108311.1 | - | 1 - 2 | 157 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cytl1
Genomic Map