Assay Details
| Gene Symbol: Noc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021315.2 | NP_067290.2 | 10 - 11 | 1387 | 104 | |
| GenBank mRNA | AB077991.1 | - | 10 - 11 | 1350 | 104 | |
| AK134051.1 | - | 10 - 11 | 1345 | 104 | ||
| AK168944.1 | - | 10 - 11 | 1388 | 104 | ||
| BC031132.1 | - | 10 - 11 | 1350 | 4954951 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Noc3l
Genomic Map