| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400003161.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003161.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003161.mm.1 |
TC1400000358.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003163.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003163.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003163.mm.1 |
TC1400000358.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003164.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003164.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003164.mm.1 |
TC1400000358.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003170.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003170.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003170.mm.1 |
TC1400000358.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003173.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003173.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003173.mm.1 |
TC1400000358.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003160.mm.1 |
NM_153800.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003160.mm.1 |
NM_153800.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003160.mm.1 |
TC1400000358.mm.1 |
MTA TC |