Assay Details
| Gene Symbol: Pax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011037.4 | NP_035167.4 | 6 - 7 | 790 | 77 | |
| GenBank mRNA | BC148232.1 | - | 6 - 7 | 794 | 40142573 | 77 |
| BC150484.1 | - | 6 - 7 | 794 | 40142579 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax2
Genomic Map