Assay Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145460.2 | NP_663435.2 | 7 - 8 | 1122 | 99 | |
| GenBank mRNA | AK031978.1 | - | 6 - 7 | 751 | 99 | |
| AK032160.1 | - | 7 - 8 | 925 | 99 | ||
| AK075924.1 | - | 6 - 7 | 748 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021709 | AK139884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021709 | AK032160.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021709 | NM_145460.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021709 | AK031978.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331903 | AK016497.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331903 | XM_133663.5 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331903 | XM_907780.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714990 | AK030946.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714990 | AK081581.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714990 | XM_001474597.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714990 | XM_001477759.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43081 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151798 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021709 | AK139884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021709 | AK032160.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021709 | NM_145460.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021709 | AK031978.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29515557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02826632 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08074515 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02826642 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151798 | - | - |
| Whole Mouse Genome G4122A | A_52_P534620 | BC019806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534620 | AK075924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534620 | AK032160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534620 | NM_145460.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534620 | AK031978.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106018 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151798 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02826642 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11806674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11806671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11806687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29515580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11539349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11359320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11125253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11663719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11239686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11539351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11135030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11483667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11807776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11125248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11135028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11483661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29515622 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10413830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413832 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447816_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426375_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413826 | - | - |
| Mouse Genome 430 2.0 Array | 1426375_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1447816_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447816_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426375_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218885_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218885_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400002931.mm.1 | NM_145460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002931.mm.1 | NM_145460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002931.mm.1 | TC1400000337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400002932.mm.1 | NM_145460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002932.mm.1 | NM_145460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002932.mm.1 | TC1400000337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400002934.mm.1 | NM_145460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002934.mm.1 | NM_145460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002934.mm.1 | TC1400000337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400002935.mm.1 | NM_145460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002935.mm.1 | NM_145460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002935.mm.1 | TC1400000337.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426375_PM_s_at | - | - |
| Mouse Expression Set 430 | 1426375_s_at | - | - |
| Mouse Expression Set 430 | 1447816_x_at | - | - |
| Mouse Expression Set 430 | 1447816_PM_x_at | - | - |
| Murine Genome U74v2 Set | 161374_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161374_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426375_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426375_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426375_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426375_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447816_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447816_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 502998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2650484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769740 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769740 | - | - |
| mouse-6 v1.1 (Illumina) | scl0218885.8_148-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0218885.8_148-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3009558 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3009558 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769740 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0218885.8_148-S | - | - |

Oxnad1
Genomic Map