Assay Details
| Gene Symbol: Lrrc71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028971.1 | NP_083247.1 | 5 - 6 | 752 | 94 | |
| GenBank mRNA | AK016993.1 | - | 5 - 6 | 680 | 94 | |
| AK029938.1 | - | 5 - 6 | 720 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc71
Genomic Map