Assay Details
| Gene Symbol: Lrrc71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028971.1 | NP_083247.1 | 6 - 7 | 821 | 76 | |
| GenBank mRNA | AK016993.1 | - | 6 - 7 | 749 | 76 | |
| AK029938.1 | - | 6 - 7 | 789 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc71
Genomic Map