Assay Details
| Gene Symbol: Mrpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC031730.1 | - | 1 - 2 | 88 | 3991689 | 79 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC031730.1 transcripts.
Set Membership: |
|

Mrpl24
Genomic Map