Assay Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133787.2 | NP_598548.1 | 11 - 12 | 1100 | 128 | |
| GenBank mRNA | AK034772.1 | - | 300 | 128 | ||
| AK088667.1 | - | 11 - 12 | 1113 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nmd3
Genomic Map