Assay Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133787.2 | NP_598548.1 | 12 - 13 | 1213 | 123 | |
| GenBank mRNA | AK034772.1 | - | 413 | 123 | ||
| AK088667.1 | - | 12 - 13 | 1226 | 123 | ||
| AK145248.1 | - | 12 - 13 | 1235 | 123 | ||
| AK166283.1 | - | 12 - 13 | 1181 | 123 | ||
| BC003842.1 | - | 12 - 13 | 1213 | 3495021 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmd3
Genomic Map