Assay Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133787.2 | NP_598548.1 | 6 - 7 | 570 | 120 | |
| GenBank mRNA | AK088667.1 | - | 6 - 7 | 583 | 120 | |
| AK143289.1 | - | 6 - 7 | 536 | 120 | ||
| AK145248.1 | - | 6 - 7 | 592 | 120 | ||
| AK166283.1 | - | 6 - 7 | 538 | 120 | ||
| BC003842.1 | - | 6 - 7 | 570 | 3495021 | 120 | |
More Information
Set Membership: |
|

Nmd3
Genomic Map