Assay Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133787.2 | NP_598548.1 | 8 - 9 | 736 | 162 | |
| GenBank mRNA | AK088667.1 | - | 8 - 9 | 749 | 162 | |
| AK145248.1 | - | 8 - 9 | 758 | 162 | ||
| AK166283.1 | - | 8 - 9 | 704 | 162 | ||
| BC003842.1 | - | 8 - 9 | 736 | 3495021 | 162 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmd3
Genomic Map