Assay Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177041.3 | NP_796015.2 | 2 - 3 | 304 | 75 | |
| GenBank mRNA | AK044501.1 | - | 313 | 75 | ||
| AK161929.1 | - | 2 - 3 | 348 | 75 | ||
| AK211873.1 | - | 1 - 2 | 171 | 75 | ||
| BC025817.1 | - | 2 - 3 | 257 | 5099127 | 75 | |
| BC046769.1 | - | 2 - 3 | 304 | 6809196 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flad1
Genomic Map