Assay Details
| Gene Symbol: Ccser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027045.1 | NP_081321.1 | 7 - 8 | 2403 | 119 | |
| NM_028407.3 | NP_082683.2 | 5 - 6 | 575 | 119 | ||
| GenBank mRNA | AK005931.1 | - | 5 - 6 | 575 | 119 | |
| AK147383.1 | - | 7 - 8 | 2398 | 119 | ||
| BC111465.1 | - | 6 - 7 | 784 | 119 | ||
| BC139090.1 | - | 6 - 7 | 2191 | 8862112 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccser2
Genomic Map