Assay Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212452.1 | NP_997617.1 | 12 - 13 | 967 | 119 | |
| GenBank mRNA | AK054129.1 | - | 515 | 119 | ||
| AK137655.1 | - | 11 - 12 | 948 | 119 | ||
| AY509975.1 | - | 12 - 13 | 967 | 119 | ||
More Information
Set Membership: |
|

Rxfp1
Genomic Map