Assay Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212452.1 | NP_997617.1 | 13 - 14 | 1043 | 144 | |
| GenBank mRNA | AK054129.1 | - | 591 | 144 | ||
| AK137655.1 | - | 12 - 13 | 1024 | 144 | ||
| AY509975.1 | - | 13 - 14 | 1043 | 144 | ||
More Information
Set Membership: |
|

Rxfp1
Genomic Map