Assay Details
| Gene Symbol: Il11ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172054.1 | NP_001165525.1 | 1 - 2 | 116 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172054.1 transcripts.
Set Membership: |
|

Il11ra1
Genomic Map