Assay Details
| Gene Symbol: Il11ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172054.1 | NP_001165525.1 | 2 - 3 | 195 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172054.1 transcripts.
Set Membership: |
|

Il11ra1
Genomic Map