Assay Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079908.2 | NP_001073377.1 | 8 - 9 | 1830 | 67 | |
| NM_001079909.2 | NP_001073378.1 | 7 - 8 | 1569 | 67 | ||
| NM_010206.3 | NP_034336.2 | 8 - 9 | 1836 | 67 | ||
| GenBank mRNA | AK028354.1 | - | 7 - 8 | 1009 | 67 | |
| AK049704.1 | - | 8 - 9 | 1796 | 67 | ||
| BC010200.1 | - | 8 - 9 | 1729 | 3967874 | 67 | |
| BC033447.1 | - | 6 - 7 | 832 | 4037899 | 67 | |
| BC053921.1 | - | 1 - 2 | 1076 | 67 | ||
| M28998.1 | - | 7 - 8 | 1134 | 67 | ||
| M33760.1 | - | 6 - 7 | 853 | 67 | ||
| M65053.1 | - | 7 - 8 | 1107 | 67 | ||
| U22324.1 | - | 7 - 8 | 1134 | 67 | ||
| U23445.1 | - | 6 - 7 | 867 | 67 | ||
| X51893.1 | - | 8 - 9 | 1191 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr1
Genomic Map