Assay Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163493.1 | NP_001156965.1 | 2 - 3 | 341 | 65 | |
| NM_146258.2 | NP_666370.3 | 2 - 3 | 341 | 65 | ||
| GenBank mRNA | AK050259.1 | - | 2 - 3 | 342 | 65 | |
| AK148227.1 | - | 2 - 3 | 300 | 65 | ||
| BC027830.1 | - | 2 - 3 | 353 | 5320966 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stard13
Genomic Map