Assay Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001492.2 | NP_001001492.2 | 1 - 2 | 65 | 136 | |
| GenBank mRNA | AK133316.1 | - | 1 - 2 | 65 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001001492.2 transcripts.
Set Membership: |
|

Lca5l
Genomic Map